
n-Tridecane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:629-50-5
:
Formula:H₃C(CH₂)₁₁CH₃
MDL Number:MFCD00008979
:184.37 g/mol
Boiling Pt:235 °C
Melting Pt:–5 °C
Density:0.755
Flash Pt:79 °C (174 °F)
Storage Temperature:Ambient
EINECS:211-093-4
UN:3295
ADR:3,III
:GHS08