
n-Tetradecane 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:629-59-4
:
Formula:H₃C(CH₂)₁₂CH₃
MDL Number:MFCD00008986
:198.39 g/mol
Boiling Pt:252…254 °C (760 mmHg)
Melting Pt:5.8 °C
Density:0.767 g/cm³
Flash Pt:99 °C
EINECS:211-096-0
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4280 to 1.4300 (20°C, 589 nm)
- GC:≥98.5 %
- HPLC:≥97.5 %