Order Entry
Export
ContactUsLinkComponent
n-Pentane ≥99%, ROTISOLV® Pestilyse®
Flammable
Health
Irritant
Environment
n-Pentane ≥99%, ROTISOLV® Pestilyse®
Catalog # ROTHT903.1
Supplier:  Roth Carl
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
n-Pentane ≥99%, ROTISOLV® Pestilyse®
Catalog # ROTHT903.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

About this item

 :
 :72.15 g/mol
Merck Index:13,07193
Boiling Pt:35…36 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.63 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
REACH:01-2119459286-30
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:109-66-0
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498