Order Entry
Export
Orders LinkContactUsLinkComponent
n-Pentane

n-Pentane

Flammable
Health
Irritant
Environment
Supplier:   Roth Carl
n-Pentane
Supplier:  Roth Carl
Certificates
n-Pentane

3 Options Available Below

Product Details & Documents
Synonyms:  Pentane
  • CAS Number:109-66-0
  • Formula:H₃C(CH₂)₃CH₃
  • MDL Number:MFCD00009498
  • Molecular Weight:72.15 g/mol
  • Merck Index:13,07193
  • Boiling Pt:35…36 °C (1013 hPa)
  • Melting Pt:–130 °C
  • Density:0.63 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • Storage Temperature:Ambient
  • EINECS:203-692-4
  • UN:1265
  • ADR:3,II
  • REACH:01-2119459286-30
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 47,70 €
    47,70 €
    1 * 1 L
       47,70 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 95,40 €
    95,40 €
    1 * 2,5 L
       95,40 €

  • 10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 321,00 €
    321,00 €
    1 * 10 L
       321,00 €