



n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:109-66-0
:
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498
:72.15 g/mol
Merck Index:13,07193
Boiling Pt:35…36 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.63 g/cm³ (20 °C)
Flash Pt:–40 °C
EINECS:203-692-4
UN:1265
ADR:3,II
REACH:01-2119459286-30
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay (on anhydrous substance):Min. 99.0 %
- Assay (C5 isomers):Min. 99.9 %
- Acidity:Max. 0.004 meq/g
- Boiling point:35 - 37 °C
- Colouration:Max. 10 APHA
- Density (20/4):0.625 - 0.632
- Aromatics (as C6H6):Max. 50 ppm
- Evaporation residue:Max. 10 ppm
- Total S (as SO4):Max. 50 ppm
- Water:Max. 100 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125)
Packaging Information
26185_N-PENTANE ANALAR NORMAPUR_BSE_REV0