



n-Pentane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109-66-0
:
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498
:72.15 g/mol
Merck Index:13,07193
Boiling Pt:35…36 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.626 g/cm³
Flash Pt:–49 °C
EINECS:203-692-4
UN:1265
ADR:3,II
REACH:01-2119459286-30
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.01 %
- GC:≥99.0 %
- UV:at 200 nm A: ≤0.699
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %