4,4,4-Trifluoro-1-phenyl-1,3-butanedione, Sigma-Aldrich®
Catalog # SIAL217042-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:216.16
:M-Clarity™ quality level = MQ200
EINECS:206-307-8
CAS Number:326-06-7
Formula:C₁₀H₇F₃O₂
MDL Number:MFCD00000425