Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4,5-bis(trifluoromethyl)aniline
2-Bromo-4,5-bis(trifluoromethyl)aniline
2-Bromo-4,5-bis(trifluoromethyl)aniline
2-Bromo-4,5-bis(trifluoromethyl)aniline
Supplier:
Thermo Scientific
Certificates
Expand image
2-Bromo-4,5-bis(trifluoromethyl)aniline
Supplier:
Thermo Scientific
About this item
CAS Number:
230295-15-5
:
Formula:
C₈H₄BrF₆N
:
308.02 g/mol