


β-Propiolactone ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:57-57-8
:
Formula:C₃H₄O₂
MDL Number:MFCD00005169
:72.06 g/mol
Merck Index:14,7820
Boiling Pt:161…163 °C
Melting Pt:–34 °C
Density:1.146
Flash Pt:70 °C (158 °F)
EINECS:200-340-1
UN:3382
ADR:6.1,I
:GHS06
:GHS08
:GHS07