Order Entry
Export
ContactUsLinkComponent
β-Propiolactone 98%, pure
Toxic
Health
β-Propiolactone 98%, pure
Certificates
undefined
Toxic
Health
β-Propiolactone 98%, pure
Certificates

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4120 to 1.4140 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Certificates
Search by Lot Number