Order Entry
Export
Orders LinkContactUsLinkComponent
n-Hexacosane 99%

n-Hexacosane 99%

n-Hexacosane 99%
Certificates
n-Hexacosane 99%

2 Options Available Below

Product Details & Documents
Synonyms:  N-hexacosane
  • CAS Number:630-01-3
  • Formula:H₃C(CH₂)₂₄CH₃
  • MDL Number:MFCD00009354
  • Molecular Weight:366.71 g/mol
  • Boiling Pt:420 °C
  • Melting Pt:56…59 °C
  • Density:0.8 g/cm³
  • Flash Pt:215 °C
  • Storage Temperature:Ambient
  • EINECS:211-124-1

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    56 to 59 °C
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 23,90 €
    23,90 €
    1 * 1 g
       23,90 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 37,10 €
    37,10 €
    1 * 5 g
       37,10 €