Order Entry
Export
ContactUsLinkComponent
 
n-Heptatriacontane
Catalog # EHERC14128000
Supplier:  EHRENSTORFER
CAS Number:  
n-Heptatriacontane
Catalog # EHERC14128000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :521.01 g/mol
Boiling Pt:504 °C (1013 hPa)
Melting Pt:77…79 °C
Storage Temperature:Ambient
EINECS:230-559-8
CAS Number:7194-84-5
Formula:H₃C(CH₂)₃₅CH₃
MDL Number:MFCD00042945