
n-Dodecane
Catalog # EHERC13060000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:170.34 g/mol
Boiling Pt:216 °C (1013 hPa)
Melting Pt:–9.6 °C
Density:0.7487 g/cm³ (20 °C)
Flash Pt:74 °C
Storage Temperature:Ambient
EINECS:203-967-9
:GHS08
CAS Number:112-40-3
Formula:H₃C(CH₂)₁₀CH₃
MDL Number:MFCD00008969