Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Propanedithiol ≥97%

1,3-Propanedithiol ≥97%

1,3-Propanedithiol ≥97%
Certificates
1,3-Propanedithiol ≥97%
1,3-Propanedithiol ≥97%

3 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 109-80-8 Molecular Formula: HS(CH2)3SH Formula Weight: 108.23 Flash Point: 63°(145°F) Refractive Index: 1.5400 Odour: Stench Melting Point: -79° Boiling Point: 168-170° Solubility: Slightly soluble in water. Miscible with alcohol, ether, chloroform, ether Beilstein Reference: 1071197 Merck Reference: 14,7801 UN No.: 3334 MDL No.: MFCD00004904

Synonyms:  1,3-Dimercaptopropane
  • CAS Number:109-80-8
  • Formula:C₃H₈S₂
  • MDL Number:MFCD00004904
  • Molecular Weight:108.23 g/mol
  • Merck Index:14,7801
  • Boiling Pt:168…170 °C
  • Melting Pt:–79 °C
  • Density:1.077
  • Flash Pt:63 °C (145 °F)
  • Storage Temperature:Ambient
  • EINECS:203-706-9
  • UN:3334
  • ADR:9,III
Product Family Options

Product Information