Order Entry
Export
Orders LinkContactUsLinkComponent
Glycerine

Glycerine

Glycerine
Certificates
Glycerine

2 Options Available Below

Product Details & Documents
Synonyms:  1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
  • CAS Number:56-81-5
  • Formula:HOCH₂CH(OH)CH₂OH
  • MDL Number:MFCD00004722
  • Molecular Weight:92.09 g/mol
  • Merck Index:13,04497
  • Boiling Pt:290 °C (1013 hPa)
  • Melting Pt:18.6 °C
  • Density:1.26 g/cm³ (20 °C)
  • Flash Pt:177 °C
  • Storage Temperature:Ambient
  • EINECS:200-289-5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L

  • Glass bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L