Order Entry
Export
Orders LinkContactUsLinkComponent
1,9-Nonanediol ≥97%

1,9-Nonanediol ≥97%

1,9-Nonanediol ≥97%
Certificates
1,9-Nonanediol ≥97%
1,9-Nonanediol ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 3937-56-2 Molecular Formula: HO(CH2)9OH Formula Weight: 160.26 Flash Point: 162°(323°F) Melting Point: 45-48° Boiling Point: 288° Beilstein Reference: 1737531 MDL No.: MFCD00002991

Synonyms:  1,9-Dihydroxy nonane, Nonane-1,9-diol
  • CAS Number:3937-56-2
  • Formula:C₉H₂₀O₂
  • MDL Number:MFCD00002991
  • Molecular Weight:160.26 g/mol
  • Boiling Pt:288 °C
  • Melting Pt:45…48 °C
  • Density:0.950
  • Flash Pt:162 °C (323 °F)
  • Storage Temperature:Ambient
  • EINECS:223-517-5
Product Family Options

Product Information