Order Entry
Export
Orders LinkContactUsLinkComponent
1,9-Nonanediol 99%

1,9-Nonanediol 99%

1,9-Nonanediol 99%
Certificates
1,9-Nonanediol 99%

2 Options Available Below

Product Details & Documents
Synonyms:  1,9-Dihydroxy nonane, Nonane-1,9-diol
  • CAS Number:3937-56-2
  • Formula:C₉H₂₀O₂
  • MDL Number:MFCD00002991
  • Molecular Weight:160.26 g/mol
  • Boiling Pt:177 °C (15 mmHg)
  • Melting Pt:44…48 °C
  • Density:0.918 g/cm³
  • Flash Pt:162 °C
  • Storage Temperature:Ambient
  • EINECS:223-517-5

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Fused crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    44 to 48 °C
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 43,70 €
    43,70 €
    1 * 25 g
       43,70 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 120,00 €
    120,00 €
    1 * 100 g
       120,00 €