Order Entry
Export
Orders LinkContactUsLinkComponent
1,7-Heptanediol ≥98%

1,7-Heptanediol ≥98%

Glass bottle for solids, 5 g
Catalog # L06384.06
1,7-Heptanediol ≥98%
Catalog # L06384.06
CAS Number:  629-30-1
1,7-Heptanediol ≥98%
1,7-Heptanediol ≥98%

Glass bottle for solids, 5 g

CAS Number:  629-30-1
Price
63,40 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 629-30-1 Molecular Formula: HO(CH2)7OH Formula Weight: 132.20 Flash Point: 127°(260°F) Refractive Index: 1.4550 Melting Point: 17-18° Boiling Point: 258-260° Beilstein Reference: 1633482 MDL No.: MFCD00002987

Synonyms:  1,7-Dihydroxy heptane
  • CAS Number:629-30-1
  • Formula:C₇H₁₆O₂
  • MDL Number:MFCD00002987
  • Molecular Weight:132.2 g/mol
  • Boiling Pt:258…260 °C
  • Melting Pt:17…18 °C
  • Density:0.953
  • Flash Pt:127 °C (260 °F)
  • Storage Temperature:Ambient
  • EINECS:211-085-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications