
(±)-1,2-Pentanediol 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5343-92-0
:
Formula:C₅H₁₂O₂
MDL Number:MFCD00010736
:104.15 g/mol
Boiling Pt:206 °C (760 mmHg)
Density:0.971 g/cm³
Flash Pt:105 °C
EINECS:226-285-3
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4380 to 1.4400 (20°C, 589 nm)
- GC:≥95.0 %