Ethylene glycol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol
CAS Number:107-21-1
:
Formula:HOCH₂CH₂OH
MDL Number:MFCD00002885
:62.07 g/mol
Merck Index:13,03832
Boiling Pt:196…198 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.115 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:116 °C
Storage Temperature:Refrigerator
EINECS:203-473-3
REACH:01-2119456816-28
:GHS07
Specifications
- Environmentally Preferable: