Order Entry
Export
Orders LinkContactUsLinkComponent
1,10-Decanediol

1,10-Decanediol

Glass bottle for solids, 0,25 g
Catalog # EHERC12095010
Supplier:   EHRENSTORFER
1,10-Decanediol
Catalog # EHERC12095010
Supplier:  EHRENSTORFER
CAS Number:  112-47-0
1,10-Decanediol

Glass bottle for solids, 0,25 g

CAS Number:  112-47-0
Price
Inquire For Price
Log in for availability
1 * 0,25 g
Product Details & Documents
Synonyms:  1,10-Dihydroxy decane
  • CAS Number:112-47-0
  • Formula:C₁₀H₂₂O₂
  • MDL Number:MFCD00004749
  • Molecular Weight:174.28 g/mol
  • Merck Index:13,02869
  • Boiling Pt:297 °C (1013 hPa)
  • Melting Pt:72 °C
  • Density:0.883 g/cm³ (80 °C)
  • Storage Temperature:Ambient
  • EINECS:203-975-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g

Specifications