Order Entry
Export
Orders LinkContactUsLinkComponent
1,10-Decanediol ≥97%

1,10-Decanediol ≥97%

1,10-Decanediol ≥97%
Certificates
1,10-Decanediol ≥97%
1,10-Decanediol ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 112-47-0 Molecular Formula: HO(CH2)10OH Formula Weight: 174.28 Flash Point: 152°(306°F) Melting Point: 71-74° Boiling Point: 297° Beilstein Reference: 1698975 Merck Reference: 14,2849 MDL No.: MFCD00004749

Synonyms:  1,10-Dihydroxy decane
  • CAS Number:112-47-0
  • Formula:C₁₀H₂₂O₂
  • MDL Number:MFCD00004749
  • Molecular Weight:174.28 g/mol
  • Merck Index:14,2849
  • Boiling Pt:297 °C
  • Melting Pt:71…74 °C
  • Density:1.080
  • Flash Pt:152 °C (306 °F)
  • Storage Temperature:Ambient
  • EINECS:203-975-2
Product Family Options

Product Information