Order Entry
Export
Orders LinkContactUsLinkComponent
1,10-Decanediol 99%

1,10-Decanediol 99%

1,10-Decanediol 99%
Certificates
1,10-Decanediol 99%

2 Options Available Below

Product Details & Documents
Synonyms:  1,10-Dihydroxy decane
  • CAS Number:112-47-0
  • Formula:C₁₀H₂₂O₂
  • MDL Number:MFCD00004749
  • Molecular Weight:174.28 g/mol
  • Merck Index:13,02869
  • Boiling Pt:297 °C (760 mmHg)
  • Melting Pt:71…75 °C
  • Density:0,883 g/cm³ (80 °C)
  • Flash Pt:152 °C
  • Storage Temperature:Ambient
  • EINECS:203-975-2

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Powder or flakes or pellets
  • Infrared spectrum:
    Conforms
  • Melting point:
    72 to 74 °C
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 47,50 €
    47,50 €
    1 * 100 g
       47,50 €

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 123,00 €
    123,00 €
    1 * 500 g
       123,00 €