Order Entry
Export
Orders LinkContactUsLinkComponent
 

Methanol

Flammable
Toxic
Health
Supplier:   Roth Carl
Methanol
Supplier:  Roth Carl
Certificates
Methanol

2 Options Available Below

Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64.6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0.7918 g/cm³ (20 °C)
  • Flash Pt:11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 41,00 €
    41,00 €
    1 * 1 L
       41,00 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 74,70 €
    74,70 €
    1 * 2,5 L
       74,70 €