Order Entry
Export
ContactUsLinkComponent
 
Flammable
Toxic
Health
Methanol
Catalog # EHERC14995000
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Flammable
Toxic
Health
Methanol
Catalog # EHERC14995000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595