3-Pentanol
Catalog # EHERC15981300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
Merck Index:13,07197
Boiling Pt:116 °C (1013 hPa)
Melting Pt:–8 °C
Density:0.82 g/cm³ (20 °C)
Flash Pt:33 °C
Storage Temperature:Ambient
EINECS:209-526-7
UN:1105
ADR:3,III
:88.15 g/mol
CAS Number:584-02-1
Formula:C₅H₁₂O
MDL Number:MFCD00004574