Order Entry
Export
Orders LinkContactUsLinkComponent
 

(±)-2-Undecanol ≥98%

(±)-2-Undecanol ≥98%
Certificates
(±)-2-Undecanol ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 1653-30-1 Molecular Formula: C11H24O Formula Weight: 172.31 Flash Point: 107°(225°F) Refractive Index: 1.4370 Odour: Mild Melting Point: 1-3° Boiling Point: 228-229° Beilstein Reference: 1719795 MDL No.: MFCD00021958

Synonyms:  Undecan-2-ol, (±)-Methyl nonyl carbinol, (±)-2-Hydroxy undecane, (±)-Nonyl methyl carbinol, 2-Undecanol
  • CAS Number:1653-30-1
  • Formula:C₁₁H₂₄O
  • MDL Number:MFCD00021958
  • Molecular Weight:172.31 g/mol
  • Boiling Pt:228…229 °C
  • Melting Pt:1…3 °C
  • Density:0.828
  • Flash Pt:107 °C (225 °F)
  • Storage Temperature:Ambient
  • EINECS:216-722-6
Product Family Options

Product Information