1-Octadecanol ≥97%
Purity: 97% CAS No.: 112-92-5 Molecular Formula: CH3(CH2)17OH Formula Weight: 270.50 Flash Point: 174°(345°F) Physical Form: Flakes or pellets Melting Point: 56-60° Boiling Point: 210°/15mm Beilstein Reference: 1362907 Merck Reference: 14,8805 MDL No.: MFCD00002823
: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
- :112-92-5
- :H₃C(CH₂)₁₆CH₂OH
- :MFCD00002823
- :270.5 g/mol
- :14,8805
- :210 °C (15 mmHg)
- :56…60 °C
- :0.805…0.815 g/cm³ (60 °C)
- :174 °C (345 °F)
- :Ambient
- :204-017-6
Purity: 97% CAS No.: 112-92-5 Molecular Formula: CH3(CH2)17OH Formula Weight: 270.50 Flash Point: 174°(345°F) Physical Form: Flakes or pellets Melting Point: 56-60° Boiling Point: 210°/15mm Beilstein Reference: 1362907 Merck Reference: 14,8805 MDL No.: MFCD00002823
: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
- :112-92-5
- :H₃C(CH₂)₁₆CH₂OH
- :MFCD00002823
- :270.5 g/mol
- :14,8805
- :210 °C (15 mmHg)
- :56…60 °C
- :0.805…0.815 g/cm³ (60 °C)
- :174 °C (345 °F)
- :Ambient
- :204-017-6