
1-Nonanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:143-08-8
:
Formula:H₃C(CH₂)₇CH₂OH
MDL Number:MFCD00002990
:144.26 g/mol
Merck Index:12,06774
Boiling Pt:215 °C (1013 hPa)
Melting Pt:–7 °C
Density:0.8419 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-583-7
UN:3082
ADR:9,III
:M-Clarity™ quality level = MQ200
Flash Pt:76 °C
:GHS07