Order Entry
Export
ContactUsLinkComponent
1-Nonanol
Irritant
1-Nonanol
Catalog # EHERC15623200
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
1-Nonanol
Catalog # EHERC15623200
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :144.26 g/mol
Merck Index:12,06774
Boiling Pt:215 °C (1013 hPa)
Melting Pt:–7 °C
Density:0.8419 g/cm³ (20 °C)
Flash Pt:76 °C
Storage Temperature:Ambient
EINECS:205-583-7
UN:3082
ADR:9,III
 :GHS07
CAS Number:143-08-8
Formula:H₃C(CH₂)₇CH₂OH
MDL Number:MFCD00002990