ContactUsLinkComponent
1-Hexanol
1-Hexanol
  PROOCHE136
 :  
undefined
1-Hexanol
  PROOCHE136
 :  
  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Seq:
    2145
 :
 :102.18 g/mol
 :13,04715
 :156 °C (1013 hPa)
 :–45 °C
 :0.8322 g/cm³ (20 °C)
 :60 °C
 :Ambient
 :203-852-3
 :2282
 :3,III
 :GHS07
 :111-27-3
 :H₃C(CH₂)₄CH₂OH
 :MFCD00002982