Order Entry
Export
Orders LinkContactUsLinkComponent
1-Hexanol ≥99%

1-Hexanol ≥99%

Flammable
Irritant
1-Hexanol ≥99%
Certificates
1-Hexanol ≥99%
1-Hexanol ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 111-27-3 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 60°(140°F) Physical Form: Clear liquid Density: 0.814 Refractive Index: 1.4180 Appearance: Colourless Melting Point: -52° Boiling Point: 157° Beilstein Reference: 969167 Merck Reference: 14,4697 UN No.: 2282 MDL No.: MFCD00002982

Synonyms:  Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane
  • CAS Number:111-27-3
  • Formula:H₃C(CH₂)₄CH₂OH
  • MDL Number:MFCD00002982
  • Molecular Weight:102.18 g/mol
  • Merck Index:14,4697
  • Boiling Pt:157 °C
  • Melting Pt:–52 °C
  • Density:0.814
  • Flash Pt:60 °C (140 °F)
  • Storage Temperature:Ambient
  • EINECS:203-852-3
  • UN:2282
  • ADR:3,III
Product Family Options

Product Information