Order Entry
Export
Orders LinkContactUsLinkComponent
Nitromethane, Sigma-Aldrich®

Nitromethane, Sigma-Aldrich®

Flammable
Irritant
Supplier:   Merck
Nitromethane, Sigma-Aldrich®
Supplier:  Merck
Certificates
Nitromethane, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
  • CAS Number:75-52-5
  • Formula:CH₃NO₂
  • MDL Number:MFCD00007400
  • Molecular Weight:61.04 g/mol
  • Merck Index:13,06644
  • Boiling Pt:101.2 °C (1013 hPa)
  • Melting Pt:–29 °C
  • Density:1.138 g/cm³ (20 °C)
  • Flash Pt:44 °C
  • Storage Temperature:Ambient
  • UN:1261
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:200-876-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 148,00 €
    148,00 €
    1 * 100 mL
       148,00 €

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 552,00 €
    552,00 €
    1 * 500 mL
       552,00 €