Order Entry
Export
Orders LinkContactUsLinkComponent
Triethylamine

Triethylamine

Glass bottle for solids, 100 mg
Catalog # PROOMM0477.07
Flammable
Corrosive
Irritant
Triethylamine
Catalog # PROOMM0477.07
CAS Number:  121-44-8
Triethylamine
Triethylamine

Glass bottle for solids, 100 mg

CAS Number:  121-44-8
Price
324,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  (Diethylamino)ethane, TEA
  • CAS Number:121-44-8
  • Formula:(C₂H₅)₃N
  • MDL Number:MFCD00009051
  • Molecular Weight:101.19 g/mol
  • Merck Index:13,09740
  • Boiling Pt:90 °C (1013 hPa)
  • Melting Pt:–115 °C
  • Density:0.729 g/cm³ (20 °C)
  • Flash Pt:–11 °C
  • Storage Temperature:Ambient
  • EINECS:204-469-4
  • UN:1296
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications