Order Entry
Export
Orders LinkContactUsLinkComponent
Triiodomethane 99+%

Triiodomethane 99+%

Irritant
Triiodomethane 99+%
Certificates
Triiodomethane 99+%

3 Options Available Below

Product Details & Documents
Synonyms:  Iodoform
  • CAS Number:75-47-8
  • Formula:CHI₃
  • MDL Number:MFCD00001069
  • Molecular Weight:393.73 g/mol
  • Merck Index:13,05055
  • Boiling Pt:218 °C (1013 hPa)
  • Melting Pt:119…122 °C
  • Density:4,1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-874-5

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Bright yellow to dark yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    119 to 122 °C
  • Loss on drying:
    ≤0.5 % (24 h) (Over silicagel)
  • Residue after ignition:
    ≤0.1 %
  • Titration:
    ≥99.0 %
  • Chloride (Cl):
    ≤110 ppm
  • Sulfate (SO4):
    ≤170 ppm
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 59,00 €
    59,00 €
    1 * 100 g
       59,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 176,00 €
    176,00 €
    1 * 500 g
       176,00 €

  • Glass bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 804,00 €
    804,00 €
    1 * 2,5 kg
       804,00 €