
1,6-Diiodohexane ≥98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:629-09-4
:
Formula:C₆H₁₂I₂
MDL Number:MFCD00001103
Boiling Pt:123…128 °C (4 mmHg)
Melting Pt:10 °C
Density:2.052
Flash Pt:>110 °C (230 °F)
:Stabilised with copper
:337.97 g/mol
EINECS:211-073-5
:GHS07
Storage Temperature:Ambient