

Trifluoroacetic acid 99.5% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76-05-1
:
Formula:C₂HF₃O₂
MDL Number:MFCD00004169
:114.02 g/mol
Merck Index:13,09754
Boiling Pt:72 °C (760 mmHg)
Melting Pt:–15 °C
Density:1.535 g/cm³
Storage Temperature:Ambient
EINECS:200-929-3
UN:2699
ADR:8,I
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.05 % (Coulometric)
- Color scale:≤10 APHA
- Titration with NaOH:99.5 to 100.5 %
- Chloride (Cl):≤10 ppm
- Fluoride (F):≤50 ppm
- Sulfate (SO4):≤10 ppm