



2,3-Dichloro-1-propanol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:616-23-9
:
Formula:C₃H₆Cl₂O
MDL Number:MFCD00040446
:128.99 g/mol
Boiling Pt:182 °C
Melting Pt:–44 °C
Density:1.361
Flash Pt:93 °C (199 °F)
EINECS:210-470-0
UN:2810
ADR:6.1,III
:GHS06
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient