


Trichloroacetic acid (TCA) ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
:76-03-9
:
:Cl₃CCOOH
:MFCD00004177
:163.39 g/mol
:13,09700
:196 °C (1013 hPa)
:54…56 °C
:1.63 g/cm³ (20 °C)
:113 °C (closed cup)
:Ambient
:200-927-2
:1839
:8,II
:GHS05
:GHS07
:GHS09
- Assay:Min. 99.5 %
- Clarity of solution:Passes test ACS
- IR Spectrum:Passes test
- Substances coloured by H2SO4:Passes test
- Melting point:Min. 57 °C
- Heavy metals (as Pb):Max. 20 ppm
- Ignition residue (SO4):Max. 0.03 %
- Insolubility in water:Max. 50 ppm
- Cl (Chloride):Max. 10 ppm
- NO3 (Nitrate):Max. 20 ppm
- PO4 (Phosphate):Max. 5 ppm
- SO4 (Sulphate):Max. 0.02 %
- Cu (Copper):Max. 5 ppm
- Fe (Iron):Max. 10 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test