Order Entry
Export
ContactUsLinkComponent
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.5 %
  • Acidity:
    Max. 0.0005 meq/g
  • Boiling point:
    39.0 - 40.5 °C
  • Colouration:
    Max. 10 APHA
  • Density (20/4):
    1.322 - 1.327
  • Substances coloured by H2SO4:
    Max. 60 APHA
  • Carbon tetrachloride:
    Max. 100 ppm
  • Chloroform:
    Max. 100 ppm
  • Ethanol:
    0.15 - 0.25 %
  • Evaporation residue:
    Max. 10 ppm
  • Free chlorine:
    Max. 0.3 ppm
  • Water:
    Max. 0.02 %
  • Al (Aluminium):
    Max. 0.05 ppm
  • B (Boron):
    Max. 0.01 ppm
  • Ba (Barium):
    Max. 0.01 ppm
  • Ca (Calcium):
    Max. 0.2 ppm
  • Cd (Cadmium):
    Max. 0.02 ppm
  • Co (Cobalt):
    Max. 0.01 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.02 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.05 ppm
  • Mn (Manganese):
    Max. 0.01 ppm
  • Ni (Nickel):
    Max. 0.02 ppm
  • Pb (Lead):
    Max. 0.05 ppm
  • Sn (Tin):
    Max. 0.05 ppm
  • Zn (Zinc):
    Max. 1 ppm

Specification Test Results