Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Dichloropropane ≥95%

1,3-Dichloropropane ≥95%
Certificates
1,3-Dichloropropane ≥95%
1,3-Dichloropropane ≥95%

2 Options Available Below

Product Details & Documents
Synonyms:  Trimethylendichloride
  • CAS Number:142-28-9
  • Formula:ClH₂CCH₂CH₂Cl
  • MDL Number:MFCD00000999
  • Molecular Weight:112.99 g/mol
  • Boiling Pt:120 °C
  • Melting Pt:–99.5 °C
  • Density:1.190
  • Flash Pt:20 °C (68 °F)
  • Storage Temperature:Ambient
  • EINECS:205-531-3
  • ADR:3,II
  •  :Density: 1.1717 g/cm³ (20 °C)
  • UN:1993
Product Family Options

Product Information