1,3-Dichloropropane 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Trimethylendichloride
- CAS Number:142-28-9
- Formula:ClH₂CCH₂CH₂Cl
- MDL Number:MFCD00000999
- Molecular Weight:112.99 g/mol
- Boiling Pt:120…122 °C (760 mmHg)
- Melting Pt:–99 °C
- Density:1.19 g/cm³
- Flash Pt:20 °C
- Storage Temperature:Ambient
- EINECS:205-531-3
- ADR:3,II
- :Density: 1.1717 g/cm³ (20 °C)
- UN:1993
2 Options Available Below
Product Details & Documents
Synonyms: Trimethylendichloride
- CAS Number:142-28-9
- Formula:ClH₂CCH₂CH₂Cl
- MDL Number:MFCD00000999
- Molecular Weight:112.99 g/mol
- Boiling Pt:120…122 °C (760 mmHg)
- Melting Pt:–99 °C
- Density:1.19 g/cm³
- Flash Pt:20 °C
- Storage Temperature:Ambient
- EINECS:205-531-3
- ADR:3,II
- :Density: 1.1717 g/cm³ (20 °C)
- UN:1993
Documents
ACRO11368
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4471 to 1.4491 (20°C, 589 nm)
- GC:≥98.5 %



