

1,2-Dichloropropane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:78-87-5
:
Formula:ClH₂CCH₂CH₂Cl
MDL Number:MFCD00000868
:112.99 g/mol
Merck Index:12,08039
Boiling Pt:96 °C (1013 hPa)
Melting Pt:–100 °C
Density:1.158 g/cm³ (20 °C)
EINECS:201-152-2
UN:1279
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:15 °C
:GHS02
:GHS07
Storage Temperature:Ambient