
Caprylic acid
Catalog # ROTH3168.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:144.21 g/mol
Merck Index:13,01771
Boiling Pt:239.7 °C (1013 hPa)
Melting Pt:16 °C
Density:0.9066 g/cm³ (20 °C)
Flash Pt:130 °C
Storage Temperature:Ambient
EINECS:204-677-5
UN:3265
ADR:8,III
:GHS05
CAS Number:124-07-2
Formula:C₈H₁₆O₂
MDL Number:MFCD00004429