Formic acid ≥98% 98+%, pure
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Aminic Acid, Methanoic acid
- CAS Number:64-18-6
- Formula:HCO₂H
- MDL Number:MFCD00003297
- Molecular Weight:46.03 g/mol
- Boiling Pt:101 °C (760 mmHg)
- Melting Pt:8 °C
- Density:1.22 g/cm³
- Flash Pt:50 °C
- Storage Temperature:Refrigerator
- EINECS:200-579-1
- ADR:8,II
- :Density: 1.22 g/cm³ (25 °C)
- UN:1779
5 Options Available Below
Product Details & Documents
Synonyms: Aminic Acid, Methanoic acid
- CAS Number:64-18-6
- Formula:HCO₂H
- MDL Number:MFCD00003297
- Molecular Weight:46.03 g/mol
- Boiling Pt:101 °C (760 mmHg)
- Melting Pt:8 °C
- Density:1.22 g/cm³
- Flash Pt:50 °C
- Storage Temperature:Refrigerator
- EINECS:200-579-1
- ADR:8,II
- :Density: 1.22 g/cm³ (25 °C)
- UN:1779
Specifications for Product Family
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3700 to 1.3720 (20°C, 589 nm)
- Color scale:≤20 APHA
- Titration with NaOH:≥98.0 %
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



