Formic acid ≥96% ≥96% ACS
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 96+% CAS No.: 64-18-6 Molecular Formula: CH2O2 Formula Weight: 46.03 Flash Point: 50°(156°F) Physical Form: Liquid Density: 1.220 Refractive Index: 1.3710 Appearance: Colourless Melting Point: 7-9° Boiling Point: 100-101° Beilstein Reference: 1209246 Merck Reference: 14,4241 UN No.: 1779 MDL No.: MFCD00003297
Synonyms: Aminic Acid, Methanoic acid
- CAS Number:64-18-6
- Formula:HCO₂H
- MDL Number:MFCD00003297
- Molecular Weight:46.03 g/mol
- Merck Index:14,4241
- Boiling Pt:100…101 °C
- Melting Pt:7…9 °C
- Density:1.220
- Flash Pt:50 °C (156 °F)
- Storage Temperature:Ambient
- UN:1779
- ADR:8,II
- :Density: 1.22 g/cm³ (25 °C)
- EINECS:200-579-1
3 Options Available Below
Product Details & Documents
Purity: 96+% CAS No.: 64-18-6 Molecular Formula: CH2O2 Formula Weight: 46.03 Flash Point: 50°(156°F) Physical Form: Liquid Density: 1.220 Refractive Index: 1.3710 Appearance: Colourless Melting Point: 7-9° Boiling Point: 100-101° Beilstein Reference: 1209246 Merck Reference: 14,4241 UN No.: 1779 MDL No.: MFCD00003297
Synonyms: Aminic Acid, Methanoic acid
- CAS Number:64-18-6
- Formula:HCO₂H
- MDL Number:MFCD00003297
- Molecular Weight:46.03 g/mol
- Merck Index:14,4241
- Boiling Pt:100…101 °C
- Melting Pt:7…9 °C
- Density:1.220
- Flash Pt:50 °C (156 °F)
- Storage Temperature:Ambient
- UN:1779
- ADR:8,II
- :Density: 1.22 g/cm³ (25 °C)
- EINECS:200-579-1



