Palmitic acid 95%
Supplier: Thermo Fisher Scientific
About this item
Purity: 95% CAS No.: 57-10-3 Molecular Formula: C16H32O2 Formula Weight: 256.43 Flash Point: 206°(402°F) Refractive Index: 1.4273 Odour: Odourless Melting Point: 62-65° Boiling Point: 215°/15mm Beilstein Reference: 607489 Merck Reference: 14,6996 MDL No.: MFCD00002747
Synonyms: Hexadecanoic acid, Hexadecylic acid
- CAS Number:57-10-3
- Formula:C₁₆H₃₂O₂
- MDL Number:MFCD00002747
- Molecular Weight:256.43 g/mol
- Merck Index:14,6996
- Boiling Pt:215 °C (15 mmHg)
- Melting Pt:62…65 °C
- Density:0.853
- Flash Pt:206 °C (402 °F)
- Storage Temperature:Ambient
- EINECS:200-312-9
3 Options Available Below
Product Details & Documents
Purity: 95% CAS No.: 57-10-3 Molecular Formula: C16H32O2 Formula Weight: 256.43 Flash Point: 206°(402°F) Refractive Index: 1.4273 Odour: Odourless Melting Point: 62-65° Boiling Point: 215°/15mm Beilstein Reference: 607489 Merck Reference: 14,6996 MDL No.: MFCD00002747
Synonyms: Hexadecanoic acid, Hexadecylic acid
- CAS Number:57-10-3
- Formula:C₁₆H₃₂O₂
- MDL Number:MFCD00002747
- Molecular Weight:256.43 g/mol
- Merck Index:14,6996
- Boiling Pt:215 °C (15 mmHg)
- Melting Pt:62…65 °C
- Density:0.853
- Flash Pt:206 °C (402 °F)
- Storage Temperature:Ambient
- EINECS:200-312-9
Specifications for Product Family
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.B20322.36 - Specifications:
More
Specifications
Cat. No.B20322.0E - Specifications:
More
Specifications
Cat. No.B20322.0C



