Palmitic acid 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Hexadecanoic acid, Hexadecylic acid
- CAS Number:57-10-3
- Formula:C₁₆H₃₂O₂
- MDL Number:MFCD00002747
- Molecular Weight:256.43 g/mol
- Merck Index:13,07064
- Boiling Pt:351.5 °C (760 mmHg)
- Melting Pt:59…63 °C
- Density:0,853 g/cm³ (20 °C)
- Flash Pt:206 °C
- Storage Temperature:Ambient
- EINECS:200-312-9
4 Options Available Below
Product Details & Documents
Synonyms: Hexadecanoic acid, Hexadecylic acid
- CAS Number:57-10-3
- Formula:C₁₆H₃₂O₂
- MDL Number:MFCD00002747
- Molecular Weight:256.43 g/mol
- Merck Index:13,07064
- Boiling Pt:351.5 °C (760 mmHg)
- Melting Pt:59…63 °C
- Density:0,853 g/cm³ (20 °C)
- Flash Pt:206 °C
- Storage Temperature:Ambient
- EINECS:200-312-9
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Powder or granules or flakes
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Acid value:215 to 233 mg KOH/g
- Iodine value:≤1 g I2/100 g
- Saponification value:216 to 224 mg KOH/g



