Capric acid 99%
: Decylic acid, Decanoic acid
- :334-48-5
- :C₁₀H₂₀O₂
- :MFCD00004441
- :172.27 g/mol
- :13,01764
- :268…270 °C (760 mmHg)
- :30…32 °C
- :0.89 g/cm³
- :150 °C
- :Ambient
- :206-376-4
: Decylic acid, Decanoic acid
- :334-48-5
- :C₁₀H₂₀O₂
- :MFCD00004441
- :172.27 g/mol
- :13,01764
- :268…270 °C (760 mmHg)
- :30…32 °C
- :0.89 g/cm³
- :150 °C
- :Ambient
- :206-376-4
- Appearance:Colorless or white to cream Crystals and/or chunks or fused solid or liquid
- Infrared spectrum:Conforms
- Melting point:29 to 34 °C
- GC:≥98.5 %
Product Family Options
- Pack typePk



