Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl acetate

Methyl acetate

Flammable
Irritant
Supplier:   Roth Carl
Methyl acetate
Supplier:  Roth Carl
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Acetic acid methyl ester
  • CAS Number:79-20-9
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00008711
  • Molecular Weight:74.08 g/mol
  • Merck Index:13,06038
  • Boiling Pt:56.9 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0.938 g/cm³ (20 °C)
  • Flash Pt:–13 °C
  • Storage Temperature:Ambient
  • EINECS:201-185-2
  • UN:1231
  • ADR:3,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 42,50 €
    42,50 €
    1 * 1 L
       42,50 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 79,90 €
    79,90 €
    1 * 2,5 L
       79,90 €